Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265981.1 | NP_001252910.1 | 900 | 110 | ||
XM_015130792.2 | XP_014986278.1 | 1008 | 110 | |||
GenBank mRNA | JV046884.1 | - | 900 | 110 |
Gene Symbol: RNF123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU346973.1 | - | 4263 | 110 | ||
JU476736.1 | - | 4804 | 110 | |||
JU476737.1 | - | 4723 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |