Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194122.2 | NP_001181051.1 | 138 | 121 | ||
XM_015124981.2 | XP_014980467.2 | 224 | 121 | |||
GenBank mRNA | JU320188.1 | - | 131 | 121 | ||
JV045506.1 | - | 132 | 121 | |||
JV489308.1 | - | 135 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |