Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152972.2 | XP_015008458.1 | 1008 | 151 | ||
GenBank mRNA | JU337308.1 | - | 730 | 151 | ||
JU478012.1 | - | 638 | 151 | |||
JU478013.1 | - | 638 | 151 | |||
JV048824.1 | - | 744 | 151 | |||
JV048825.1 | - | 744 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |