Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097187.4 | XP_001097187.1 | 430 | 131 | ||
XM_015115483.2 | XP_014970969.1 | 424 | 131 | |||
GenBank mRNA | JU326704.1 | - | 373 | 131 | ||
JV710422.1 | - | 376 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |