Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086222.4 | XP_001086222.2 | 479 | 146 | ||
XM_028848904.1 | XP_028704737.1 | 460 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |