Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSL1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106758.4 | XP_001106758.2 | 596 | 84 | ||
XM_015125631.2 | XP_014981117.2 | 520 | 84 | |||
GenBank mRNA | JU484284.1 | - | 566 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |