Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148395.2 | XP_015003881.1 | 7693 | 150 | ||
XM_015148396.2 | XP_015003882.1 | 1917 | 150 | |||
XM_015148397.2 | XP_015003883.1 | 1757 | 150 | |||
XM_028826975.1 | XP_028682808.1 | 1761 | 150 | |||
XM_028826976.1 | XP_028682809.1 | 1906 | 150 | |||
GenBank mRNA | JU472197.1 | - | 1703 | 150 | ||
JU472198.1 | - | 1703 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |