Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116456.2 | XP_014971942.2 | 957 | 75 | ||
XM_015117111.2 | XP_014972597.1 | 998 | 75 | |||
GenBank mRNA | JU334810.1 | - | 847 | 75 | ||
JU475835.1 | - | 867 | 75 | |||
JV600290.1 | - | 899 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |