Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194753.2 | NP_001181682.1 | 449 | 115 | ||
XM_001114790.4 | XP_001114790.1 | 503 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |