Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261280.1 | NP_001248209.1 | 725 | 165 | ||
XM_015138149.2 | XP_014993635.1 | 4859 | 165 | |||
XM_015138150.2 | XP_014993636.1 | 610 | 165 | |||
XM_015138151.2 | XP_014993637.1 | 648 | 165 | |||
GenBank mRNA | JU334842.1 | - | 638 | 165 | ||
JU472395.1 | - | 738 | 165 | |||
JV047272.1 | - | 725 | 165 | |||
JV686101.1 | - | 885 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |