Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101773.4 | XP_001101773.1 | 806 | 61 | ||
XM_002801029.3 | XP_002801075.2 | 678 | 61 | |||
XM_015122508.2 | XP_014977994.1 | 623 | 61 | |||
XM_028839983.1 | XP_028695816.1 | 766 | 61 | |||
XM_028839985.1 | XP_028695818.1 | 583 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |