Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148410.2 | XP_015003896.1 | 2208 | 95 | ||
XM_015148412.2 | XP_015003898.1 | 2076 | 95 | |||
XM_028826981.1 | XP_028682814.1 | 3213 | 95 | |||
GenBank mRNA | JU335112.1 | - | 1952 | 95 | ||
JU470643.1 | - | 1958 | 95 | |||
JV047453.1 | - | 1959 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |