Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113270.4 | XP_001113270.1 | 1259 | 100 | ||
GenBank mRNA | JV635688.1 | - | 100 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |