Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149557.2 | XP_015005043.2 | 2159 | 125 | ||
XM_015149559.2 | XP_015005045.2 | 2229 | 125 | |||
XM_028828148.1 | XP_028683981.1 | 2294 | 125 | |||
XM_028828149.1 | XP_028683982.1 | 2185 | 125 | |||
XM_028828150.1 | XP_028683983.1 | 2694 | 125 | |||
XM_028828151.1 | XP_028683984.1 | 2676 | 125 | |||
XM_028828154.1 | XP_028683987.1 | 2628 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |