Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194895.2 | NP_001181824.1 | 1573 | 121 | ||
XM_001089352.4 | XP_001089352.2 | 1767 | 121 | |||
XM_015143759.2 | XP_014999245.1 | 1395 | 121 | |||
GenBank mRNA | JU335416.1 | - | 1645 | 121 | ||
JU471388.1 | - | 1554 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |