Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093783.4 | XP_001093783.2 | 1573 | 125 | ||
XM_015149581.2 | XP_015005067.2 | 1595 | 125 | |||
XM_015149582.2 | XP_015005068.2 | 1566 | 125 | |||
XM_015149583.2 | XP_015005069.2 | 1504 | 125 | |||
XM_015149584.2 | XP_015005070.2 | 1437 | 125 | |||
XM_015149586.2 | XP_015005072.2 | 1655 | 125 | |||
XM_028828134.1 | XP_028683967.1 | 1259 | 125 | |||
XM_028828135.1 | XP_028683968.1 | 1259 | 125 | |||
XM_028828136.1 | XP_028683969.1 | 1260 | 125 | |||
XM_028828137.1 | XP_028683970.1 | 1260 | 125 | |||
XM_028828138.1 | XP_028683971.1 | 1301 | 125 | |||
XM_028828139.1 | XP_028683972.1 | 1429 | 125 | |||
XM_028828140.1 | XP_028683973.1 | 1347 | 125 | |||
GenBank mRNA | JU335449.1 | - | 1574 | 125 | ||
JU476393.1 | - | 1580 | 125 | |||
JV587255.1 | - | 853 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |