Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266383.1 | NP_001253312.1 | 1407 | 153 | ||
GenBank mRNA | JU320654.1 | - | 1407 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |