Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116444.2 | XP_014971930.2 | 478 | 104 | ||
GenBank mRNA | JV690484.1 | - | 391 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |