Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194219.2 | NP_001181148.1 | 749 | 153 | ||
XM_001102761.3 | XP_001102761.1 | 595 | 153 | |||
XM_002799644.3 | XP_002799690.1 | 514 | 153 | |||
XM_015114743.2 | XP_014970229.1 | 683 | 153 | |||
GenBank mRNA | JU335504.1 | - | 627 | 153 | ||
JU476442.1 | - | 667 | 153 | |||
JV047683.1 | - | 685 | 153 | |||
JV690157.1 | - | 691 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |