Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086528.4 | XP_001086528.1 | 2129 | 103 | ||
XM_015145588.2 | XP_015001074.1 | 2151 | 103 | |||
XM_015145589.2 | XP_015001075.1 | 2129 | 103 | |||
XM_015145590.2 | XP_015001076.1 | 2129 | 103 | |||
XM_015145591.2 | XP_015001077.1 | 2129 | 103 | |||
XM_015145592.2 | XP_015001078.1 | 2129 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |