Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134545.2 | XP_014990031.2 | 1690 | 90 | ||
XM_028845622.1 | XP_028701455.1 | 1443 | 90 | |||
XM_028845623.1 | XP_028701456.1 | 1261 | 90 | |||
XM_028845624.1 | XP_028701457.1 | 1443 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |