Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261147.1 | NP_001248076.1 | 290 | 150 | ||
XM_015114052.2 | XP_014969538.1 | 334 | 150 | |||
GenBank mRNA | JU323433.1 | - | 290 | 150 | ||
JV490267.1 | - | 168 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |