Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261578.2 | NP_001248507.1 | 199 | 140 | ||
XM_015142938.2 | XP_014998424.1 | 391 | 140 | |||
GenBank mRNA | JU335969.1 | - | 195 | 140 | ||
JU476852.1 | - | 199 | 140 | |||
JV659122.1 | - | 199 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |