Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H7orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133262.2 | XP_014988748.2 | 719 | 150 | ||
GenBank mRNA | JU328451.1 | - | 118 | 150 | ||
JU328452.1 | - | 118 | 150 | |||
JU493253.1 | - | 188 | 150 | |||
JV723167.1 | - | 118 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |