Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265791.1 | NP_001252720.1 | 679 | 94 | ||
XM_028826336.1 | XP_028682169.1 | 426 | 94 | |||
GenBank mRNA | JU328456.1 | - | 498 | 94 | ||
JV628546.1 | - | 680 | 94 | |||
JV636373.1 | - | 679 | 94 | |||
JV723250.1 | - | 479 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |