Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112368.4 | XP_001112368.3 | 280 | 63 | ||
XM_028831702.1 | XP_028687535.1 | 280 | 63 | |||
GenBank mRNA | JV723624.1 | - | 63 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |