Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089856.4 | XP_001089856.3 | 513 | 131 | ||
XM_028837907.1 | XP_028693740.1 | 513 | 131 | |||
GenBank mRNA | JU336436.1 | - | 399 | 131 | ||
JV482278.1 | - | 318 | 131 | |||
JV565189.1 | - | 394 | 131 | |||
JV723685.1 | - | 456 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |