Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148216.2 | XP_015003702.2 | 1930 | 154 | ||
XM_015148217.2 | XP_015003703.2 | 1930 | 154 | |||
XM_028826883.1 | XP_028682716.1 | 1926 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |