Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRPEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261551.1 | NP_001248480.1 | 251 | 81 | ||
XM_028848093.1 | XP_028703926.1 | 410 | 81 | |||
XM_028848094.1 | XP_028703927.1 | 410 | 81 | |||
GenBank mRNA | JU320862.1 | - | 251 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |