Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107356.4 | XP_001107356.2 | 1519 | 92 | ||
XM_015148892.2 | XP_015004378.1 | 4988 | 92 | |||
XM_028828473.1 | XP_028684306.1 | 1594 | 92 | |||
XM_028828474.1 | XP_028684307.1 | 1560 | 92 | |||
GenBank mRNA | JV072377.1 | - | 1368 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |