Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083260.4 | XP_001083260.1 | 340 | 109 | ||
GenBank mRNA | JV717138.1 | - | 299 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |