Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM219A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193839.2 | NP_001180768.1 | 649 | 66 | ||
XM_002800103.3 | XP_002800149.1 | 633 | 66 | |||
XM_015117356.2 | XP_014972842.1 | 684 | 66 | |||
XM_015117357.2 | XP_014972843.1 | 633 | 66 | |||
GenBank mRNA | JU322134.1 | - | 602 | 66 | ||
JU322135.1 | - | 602 | 66 | |||
JU322219.1 | - | 653 | 66 | |||
JU472014.1 | - | 585 | 66 | |||
JV601564.1 | - | 653 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |