Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193985.1 | NP_001180914.1 | 666 | 93 | ||
GenBank mRNA | JV665604.1 | - | 452 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |