Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257889.1 | NP_001244818.1 | 1239 | 129 | ||
XM_015110633.2 | XP_014966119.2 | 1208 | 129 | |||
XM_028830481.1 | XP_028686314.1 | 1189 | 129 | |||
XM_028830482.1 | XP_028686315.1 | 1136 | 129 | |||
XM_028830483.1 | XP_028686316.1 | 1074 | 129 | |||
XM_028830484.1 | XP_028686317.1 | 1016 | 129 | |||
GenBank mRNA | JU335979.1 | - | 1239 | 129 | ||
JU335980.1 | - | 1181 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |