Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112797.4 | XP_001112797.1 | 772 | 154 | ||
XM_001112827.4 | XP_001112827.1 | 945 | 154 | |||
XM_001112859.4 | XP_001112859.1 | 1018 | 154 | |||
XM_015118564.2 | XP_014974050.1 | 841 | 154 | |||
XM_015118565.2 | XP_014974051.1 | 711 | 154 | |||
XM_015118566.2 | XP_014974052.2 | 788 | 154 | |||
XM_015118567.2 | XP_014974053.1 | 638 | 154 | |||
XM_028836000.1 | XP_028691833.1 | 464 | 154 | |||
GenBank mRNA | JU325022.1 | - | 138 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |