Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082541.4 | XP_001082541.3 | 664 | 117 | ||
XM_015146456.2 | XP_015001942.2 | 861 | 117 | |||
XM_028825985.1 | XP_028681818.1 | 1390 | 117 | |||
XM_028825986.1 | XP_028681819.1 | 697 | 117 | |||
XM_028825987.1 | XP_028681820.1 | 676 | 117 | |||
XM_028825988.1 | XP_028681821.1 | 1846 | 117 | |||
XM_028825989.1 | XP_028681822.1 | 445 | 117 | |||
GenBank mRNA | JU483835.1 | - | 227 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |