Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128627.2 | XP_014984113.2 | 147 | 52 | ||
XM_028841556.1 | XP_028697389.1 | 147 | 52 | |||
XM_028841560.1 | XP_028697393.1 | 158 | 52 | |||
XM_028841566.1 | XP_028697399.1 | 147 | 52 | |||
GenBank mRNA | JV642160.1 | - | 105 | 52 |
Gene Symbol: INTS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU488912.1 | - | 3068 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |