Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134625.2 | XP_014990111.2 | 1837 | 131 | ||
XM_015134636.2 | XP_014990122.2 | 1876 | 131 | |||
XM_015134643.2 | XP_014990129.2 | 1871 | 131 | |||
XM_028836364.1 | XP_028692197.1 | 1876 | 131 | |||
XM_028836369.1 | XP_028692202.1 | 1883 | 131 | |||
XM_028836372.1 | XP_028692205.1 | 1885 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |