Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100303.4 | XP_001100303.3 | 1802 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Macaca mulatta |