Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194539.2 | NP_001181468.2 | 748 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |