Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129891.2 | XP_014985377.1 | 560 | 133 | ||
XM_015129892.2 | XP_014985378.1 | 513 | 133 | |||
XM_015129893.2 | XP_014985379.1 | 454 | 133 | |||
XM_015129894.2 | XP_014985380.1 | 560 | 133 | |||
XM_028844610.1 | XP_028700443.1 | 252 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |