Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028845920.1 | XP_028701753.1 | 428 | 112 | ||
GenBank mRNA | JU333412.1 | - | 283 | 112 | ||
JU474698.1 | - | 290 | 112 | |||
JV608993.1 | - | 292 | 112 | |||
JV645327.1 | - | 297 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |