Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194699.2 | NP_001181628.1 | 227 | 101 | ||
XM_015124415.2 | XP_014979901.1 | 292 | 101 | |||
GenBank mRNA | JU322500.1 | - | 273 | 101 | ||
JU484832.1 | - | 358 | 101 | |||
JV045277.1 | - | 267 | 101 | |||
JV579089.1 | - | 276 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |