Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194846.3 | NP_001181775.2 | 192 | 89 | ||
XM_001111791.4 | XP_001111791.2 | 238 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |