Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265809.1 | NP_001252738.1 | 187 | 95 | ||
XM_015118755.2 | XP_014974241.1 | 452 | 95 | |||
XM_015118761.2 | XP_014974247.1 | 462 | 95 | |||
XM_028843268.1 | XP_028699101.1 | 508 | 95 | |||
XM_028843282.1 | XP_028699115.1 | 358 | 95 | |||
GenBank mRNA | JV635777.1 | - | 187 | 95 | ||
JV672885.1 | - | 367 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |