Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115536.2 | XP_014971022.2 | 533 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |