Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123900.2 | XP_014979386.1 | 1367 | 133 | ||
XM_015123909.2 | XP_014979395.1 | 1508 | 133 | |||
XM_015123916.2 | XP_014979402.1 | 1414 | 133 | |||
XM_015123920.2 | XP_014979406.1 | 1274 | 133 | |||
XM_028850547.1 | XP_028706380.1 | 1499 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |