Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120493.2 | XP_014975979.2 | 473 | 83 | ||
XM_015120494.2 | XP_014975980.1 | 473 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |