Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193479.2 | NP_001180408.1 | 593 | 123 | ||
GenBank mRNA | JU474687.1 | - | 571 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |