Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101035.4 | XP_001101035.2 | 1259 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |